missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichloropyridine, 99%, Thermo Scientific Chemicals
40.60 € - 1067.00 €
Chemical Identifiers
CAS | 2402-77-9 |
---|---|
Molecular Formula | C5H3Cl2N |
Molecular Weight (g/mol) | 147.99 |
MDL Number | MFCD00006229 |
InChI Key | MAKFMOSBBNKPMS-UHFFFAOYSA-N |
Synonym | 2,3-dichloro-pyridine, pyridine, 2,3-dichloro, dichloropyridine, pyridine, dichloro, ccris 1726, 2,3-dichloro pyridine, pubchem1198, pyridine,3-dichloro, 2.3-dichloro-pyridine, acmc-209g7xShow More |
PubChem CID | 16988 |
IUPAC Name | 2,3-dichloropyridine |
SMILES | ClC1=CC=CN=C1Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10152570
|
Thermo Scientific Acros
113720025 |
2.5 g | Glass bottle | N/A | N/A | N/A | |||||
10224380
|
Thermo Scientific Acros
113720100 |
10 g | Glass bottle |
44.82 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10395980
|
Thermo Scientific
113720500 |
50 g | Glass bottle |
149.75 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11926291
|
Thermo Scientific Acros
113722500 |
250 g | Glass Bottle | N/A | N/A | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2402-77-9 | |
147.99 | |
MAKFMOSBBNKPMS-UHFFFAOYSA-N | |
16988 | |
ClC1=CC=CN=C1Cl |
C5H3Cl2N | |
MFCD00006229 | |
2,3-dichloro-pyridine, pyridine, 2,3-dichloro, dichloropyridine, pyridine, dichloro, ccris 1726, 2,3-dichloro pyridine, pubchem1198, pyridine,3-dichloro, 2.3-dichloro-pyridine, acmc-209g7xShow More | |
2,3-dichloropyridine |
Specifications
2402-77-9 | |
White to Beige | |
99% | |
(C5H3N)Cl2 | |
2,3-dichloro-pyridine, pyridine, 2,3-dichloro, dichloropyridine, pyridine, dichloro, ccris 1726, 2,3-dichloro pyridine, pubchem1198, pyridine,3-dichloro, 2.3-dichloro-pyridine, acmc-209g7x | |
ClC1=CC=CN=C1Cl | |
147.99 | |
147.99 | |
Crystalline Powder |
67°C to 70°C | |
Authentic | |
C5H3Cl2N | |
MFCD00006229 | |
MAKFMOSBBNKPMS-UHFFFAOYSA-N | |
2,3-dichloropyridine | |
16988 | |
≥98.5% | |
2, 3-Dichloropyridine |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,3-Dichloropyridine, 99%, Thermo Scientific Chemicals