missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2',3'-Dideoxyinosine, 98%

Product Code. 15406609
Click to view available options
Quantity:
250 mg
500 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2?,3?-Dideoxyinosine is a potent anti-retroviral agent. It is most effective in combination therapy for the treatment of HIV and related lymphoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,3′-Dideoxyinosine is a potent anti-retroviral agent. It is most effective in combination therapy for the treatment of HIV and related lymphoma.

Solubility
Soluble in DMSO or methanol

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69655-05-6
Molecular Formula C10H12N4O3
Molecular Weight (g/mol) 236.23
MDL Number MFCD00077728
InChI Key BXZVVICBKDXVGW-JHPDDGAFNA-N
Synonym didanosine, 2',3'-dideoxyinosine, dideoxyinosine, videx, videx ec, ddino, inosine, 2',3'-dideoxy, didanosina, didanosinum, ddi
PubChem CID 50599
ChEBI CHEBI:490877
IUPAC Name 9-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES OC[C@@H]1CC[C@@H](O1)N1C=NC2=C1N=CNC2=O

Specifications

Color White
Quantity 250 mg
Beilstein 3619529
Merck Index 143098
Solubility Information Soluble in DMSO or methanol
Formula Weight 236.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2',3'-Dideoxyinosine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'