missing translation for 'onlineSavingsMsg'
Learn More
2,3-Difluoro-4-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals Product Code.: 15424137

2,3-Difluoro-4-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 15424137
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15424137

Brand: Thermo Scientific Alfa Aesar H31538.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 676500-39-3
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.10
MDL Number MFCD08235194
InChI Key KTGPDDPEVRJSFY-UHFFFAOYSA-N
PubChem CID 23107063
IUPAC Name 2,3-difluoro-4-hydroxybenzaldehyde
SMILES OC1=C(F)C(F)=C(C=O)C=C1

Specifications

Melting Point 140°C to 144°C
Quantity 250 mg
Sensitivity Air Sensitive
Synonym 2,3-difluoro-4-hydroxy-benzaldehyde, benzaldehyde, 2,3-difluoro-4-hydroxy, pubchem10121, benzaldehyde,2,3-difluoro-4-hydroxy, 2,3-bis fluoranyl-4-oxidanyl-benzaldehyde, benzaldehyde, 2,3-difluoro-4-hydroxy-9ci
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material 2,3-Difluoro-4-hydroxybenzaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10155955 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH31538.MD,H31538.03
Lot Number10205330 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH31538.MD,H31538.03
Lot Number10217805 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH31538.MD,H31538.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3-Difluoro-4-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.