missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-methylphenol, 97%, Thermo Scientific™

52.00 € - 174.00 €

Chemical Identifiers

CAS 261763-45-5
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.12
MDL Number MFCD01631651
InChI Key NVYIUITYVMVHJM-UHFFFAOYSA-N
Synonym phenol, 2,3-difluoro-4-methyl, acmc-20ajxd, 2,3-difluoro-4-methyl phenol, 2,3-difluoro-4-methyl-phenol, phenol, 2,3-difluoro-4-methyl-9ci
PubChem CID 2774151
IUPAC Name 2,3-difluoro-4-methylphenol
SMILES CC1=C(F)C(F)=C(O)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11386949
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H26821.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11396949
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H26821.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

261763-45-5
144.12
NVYIUITYVMVHJM-UHFFFAOYSA-N
2774151
CC1=C(F)C(F)=C(O)C=C1
C7H6F2O
MFCD01631651
phenol, 2,3-difluoro-4-methyl, acmc-20ajxd, 2,3-difluoro-4-methyl phenol, 2,3-difluoro-4-methyl-phenol, phenol, 2,3-difluoro-4-methyl-9ci
2,3-difluoro-4-methylphenol

Specifications

261763-45-5
C7H6F2O
UN3261
NVYIUITYVMVHJM-UHFFFAOYSA-N
2,3-difluoro-4-methylphenol
2774151
97%
35°C to 38°C
MFCD01631651
phenol, 2,3-difluoro-4-methyl, acmc-20ajxd, 2,3-difluoro-4-methyl phenol, 2,3-difluoro-4-methyl-phenol, phenol, 2,3-difluoro-4-methyl-9ci
CC1=C(F)C(F)=C(O)C=C1
144.12
144.12
2,3-Difluoro-4-methylphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications