missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorophenol, 98+%, Thermo Scientific Chemicals

Product Code. 11468806
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6418-38-8
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.094
MDL Number MFCD00010262
InChI Key RPEPGIOVXBBUMJ-UHFFFAOYSA-N
Synonym phenol, 2,3-difluoro, phenol, difluoro, 2,3-difluoro phenol, difluorophenol, difluoro-phenol, 2.3-difluorophenol, 2,3-difluoro-phenol, pubchem1494, acmc-1b6pt, 2,3-bis fluoranyl phenol
PubChem CID 80879
IUPAC Name 2,3-difluorophenol
SMILES C1=CC(=C(C(=C1)F)F)O

Specifications

Melting Point 34°C to 38°C
Boiling Point 53°C to 55°C (25 mmHg)
Flash Point 56°C (132°F)
Odor Phenol-like
Quantity 5 g
UN Number UN2921
Beilstein 2082265
Formula Weight 130.09
Percent Purity ≥98%
Chemical Name or Material 2,3-Difluorophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.