missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1H-indol-2-ylmethanol, Thermo Scientific™

Product Code. 10157042
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 27640-31-9
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
InChI Key GRPOFAKYHPAXNP-UHFFFAOYSA-N
Synonym indolin-2-ylmethanol, 2,3-dihydro-1h-indol-2-yl-methanol, 1h-indole-2-methanol, 2,3-dihydro, 2,3-dihydro-1h-indol-2-yl methanol, 2-hydroxymethylindoline, acmc-209gye, 2-hydroxymethyl-indoline, indolin-2-ylmethan-1-ol, rs-2-hydroxymethylindoline, +-2-hydroxymethyl-indoline
PubChem CID 2794664
IUPAC Name 2,3-dihydro-1H-indol-2-ylmethanol
SMILES C1C(NC2=CC=CC=C21)CO

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 149.19
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydro-1H-indol-2-ylmethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'