missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-7-azaindole, ≥97%, Thermo Scientific Chemicals

Product Code. 15411829
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It can be used as pharmaceutical raw material and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10592-27-5
Molecular Formula C7H8N2
Molecular Weight (g/mol) 120.155
MDL Number MFCD06659751
InChI Key ZFFYPGZDXUPKNK-UHFFFAOYSA-N
Synonym 2,3-dihydro-1h-pyrrolo 2,3-b pyridine, 7-azaindoline, 2,3-dihydro-7-azaindole, 1h,2h,3h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 2,3-dihydro, 2,3-dihydro-1h-pyrrolo 2,3-b pyridine 7-azaindoline, pubchem18022, acmc-2098hy, ksc492q5f
PubChem CID 10011889
IUPAC Name 2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
SMILES C1CNC2=C1C=CC=N2

Specifications

Melting Point 78°C to 83°C
Color White
Quantity 5 g
Beilstein 116366
Solubility Information Slightly soluble in water.
Formula Weight 120.15
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 2,3-Dihydro-7-azaindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'