missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carboxylic acid, ≥97%, Thermo Scientific™

20.90 € - 77.30 €

Chemical Identifiers

CAS 260063-21-6
Molecular Formula C7H6O4S
Molecular Weight (g/mol) 186.181
MDL Number MFCD07801102
InChI Key DCCRIIFBJZOPAV-UHFFFAOYSA-N
Synonym 2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxylic acid, 2,3-dihydro-thieno 3,4-b 1,4 dioxine-5-carboxylic acid, 2h,3h-thieno 3,4-b 1,4 dioxine-5-carboxylic acid, thieno 3,4-b-1,4-dioxin-5-carboxylicacid, 2,3-dihydro, 2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carboxylic acid, 3,4-ethylenedioxythiophene carboxylic acid, thieno 3,4-b-1,4-dioxin-5-carboxylic acid, 2,3-dihydro
PubChem CID 6483880
IUPAC Name 2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxylic acid
SMILES C1COC2=C(SC=C2O1)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10641475
View Documents Promotion Details
Thermo Scientific Maybridge
CC47801CB
250 mg Amber Glass Bottle
20.90 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10235624
View Documents Promotion Details
Thermo Scientific Maybridge
CC47801DA
1 g Amber Glass Bottle
77.30 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

260063-21-6
186.181
DCCRIIFBJZOPAV-UHFFFAOYSA-N
6483880
C1COC2=C(SC=C2O1)C(=O)O
C7H6O4S
MFCD07801102
2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxylic acid, 2,3-dihydro-thieno 3,4-b 1,4 dioxine-5-carboxylic acid, 2h,3h-thieno 3,4-b 1,4 dioxine-5-carboxylic acid, thieno 3,4-b-1,4-dioxin-5-carboxylicacid, 2,3-dihydro, 2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carboxylic acid, 3,4-ethylenedioxythiophene carboxylic acid, thieno 3,4-b-1,4-dioxin-5-carboxylic acid, 2,3-dihydro
2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxylic acid

Specifications

260063-21-6
100.0
C7H6O4S
2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxylic acid, 2,3-dihydro-thieno 3,4-b 1,4 dioxine-5-carboxylic acid, 2h,3h-thieno 3,4-b 1,4 dioxine-5-carboxylic acid, thieno 3,4-b-1,4-dioxin-5-carboxylicacid, 2,3-dihydro, 2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carboxylic acid, 3,4-ethylenedioxythiophene carboxylic acid, thieno 3,4-b-1,4-dioxin-5-carboxylic acid, 2,3-dihydro
C1COC2=C(SC=C2O1)C(=O)O
186.181
186.19
2,3-Dihydrothieno[3,4-b][1,4]dioxine-5-carboxylic acid
96.5
97%
MFCD07801102
DCCRIIFBJZOPAV-UHFFFAOYSA-N
2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carboxylic acid
6483880
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications