Learn More
2,3-Dimethyl-1,3-butadiene, 98%, stabilized with BHT, Thermo Scientific Chemicals
34.97 € - 291.45 €
Chemical Identifiers
CAS | 513-81-5 |
---|---|
Molecular Formula | C6H10 |
Molecular Weight (g/mol) | 82.15 |
MDL Number | MFCD00008595 |
InChI Key | SDJHPPZKZZWAKF-UHFFFAOYSA-N |
Synonym | 2,3-dimethyl-1,3-butadiene, biisopropenyl, 2,3-dimethylbutadiene, diisopropenyl, 2,3-dimethylenebutane, 1,3-butadiene, 2,3-dimethyl, unii-61tuu25hco, 61tuu25hco, 2,3-dimethyl-buta-1,3-diene, 2,3-diene |
PubChem CID | 10566 |
IUPAC Name | 2,3-dimethylbuta-1,3-diene |
SMILES | CC(=C)C(C)=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10239653
|
Thermo Scientific Acros
163430050 |
5 g | Glass bottle |
34.97 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10065823
|
Thermo Scientific Acros
163430250 |
25 g | Glass bottle |
95.68 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10025873
|
Thermo Scientific Acros
163431000 |
100 g | Glass bottle |
291.45 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
513-81-5 | |
82.15 | |
SDJHPPZKZZWAKF-UHFFFAOYSA-N | |
10566 | |
CC(=C)C(C)=C |
C6H10 | |
MFCD00008595 | |
2,3-dimethyl-1,3-butadiene, biisopropenyl, 2,3-dimethylbutadiene, diisopropenyl, 2,3-dimethylenebutane, 1,3-butadiene, 2,3-dimethyl, unii-61tuu25hco, 61tuu25hco, 2,3-dimethyl-buta-1,3-diene, 2,3-diene | |
2,3-dimethylbuta-1,3-diene |
Specifications
513-81-5 | |
Colorless | |
68.0°C to 69.0°C | |
Authentic | |
C6H10 | |
1.438 | |
01, III, 991 | |
15, 3260 | |
2,3-dimethyl-1,3-butadiene, biisopropenyl, 2,3-dimethylbutadiene, diisopropenyl, 2,3-dimethylenebutane, 1,3-butadiene, 2,3-dimethyl, unii-61tuu25hco, 61tuu25hco, 2,3-dimethyl-buta-1,3-diene, 2,3-diene | |
CC(=C)C(C)=C | |
100ppm BHT | |
10566 | |
98% | |
2, 3-Dimethyl-1, 3-butadiene, Stabilized with BHT |
-76.0°C | |
0.7260g/mL | |
−20°C | |
97.5% min. (GC) | |
H2C=C(CH3)C(CH3)=CH2 | |
MFCD00008595 | |
01,276; 13,45 | |
0.726 | |
SDJHPPZKZZWAKF-UHFFFAOYSA-N | |
2,3-dimethylbuta-1,3-diene | |
82.15 | |
82.13 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-172-
- 2,3-Dimethyl-1,3-butadiene
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
Supplemental information
- MIXTURE LIST-Contains : 2,3-dimethyl-1,3-Butadiene
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.