missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Dimethylamino)propoxy]aniline, 97%, Thermo Scientific™

Product Code. 10193174
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1134-76-5
Molecular Formula C11H18N2O
Molecular Weight (g/mol) 194.278
MDL Number MFCD08687610
InChI Key XGTIDBFMHOJXSS-UHFFFAOYSA-N
Synonym 2-3-dimethylamino propoxy aniline, benzenamine, 2-3-dimethylamino propoxy, 2-3-dimethylaminopropoxy aniline, 3-2-aminophenoxy propyl dimethylamine, 2-3-dimethylamino-propoxy-phenylamine
PubChem CID 18672712
IUPAC Name 2-[3-(dimethylamino)propoxy]aniline
SMILES CN(C)CCCOC1=CC=CC=C1N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 194.28
Percent Purity 97%
Chemical Name or Material 2-[3-(Dimethylamino)propoxy]aniline
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'