missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylhydroquinone, 97%, Thermo Scientific Chemicals

Product Code. 11914681
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 608-43-5
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00009997
InChI Key BXJGUBZTZWCMEX-UHFFFAOYSA-N
Synonym 2,3-dimethylhydroquinone, o-xylene-3,6-diol, 2,3-xylohydroquinone, o-xylohydroquinone, 1,4-benzenediol, 2,3-dimethyl, unii-f0l7hg609j, 1,4-dihydroxy-2,3-dimethylbenzene, 3,6-dihydroxy-o-xylene, 2,3-dimethyl-1,4-benzenediol, 2,3-dimethyl hydroquinone
PubChem CID 69100
IUPAC Name 2,3-dimethylbenzene-1,4-diol
SMILES CC1=C(C)C(O)=CC=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 223.0°C to 225.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 138.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 3-Dimethylhydroquinone
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.