missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylpiperazine, cis + trans, 95%, Thermo Scientific Chemicals

102.00 € - 285.42 €

Chemical Identifiers

CAS 84468-52-0
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD07373397
InChI Key COWPTMLRSANSMQ-UHFFFAOYSA-N
Synonym 2,3-dimethyl-piperazine, piperazine, 2,3-dimethyl, 2,3-dimethylpiperazine, 2,3-dimethylpiperazine, cis + trans
PubChem CID 522814
IUPAC Name 2,3-dimethylpiperazine
SMILES CC1C(NCCN1)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15413437
View Documents
Thermo Scientific Alfa Aesar
H26252.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11346719
View Documents
Thermo Scientific Alfa Aesar
H26252.03
1 g
285.42 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation

Chemische Identifikatoren

84468-52-0
114.192
COWPTMLRSANSMQ-UHFFFAOYSA-N
522814
CC1C(NCCN1)C
C6H14N2
MFCD07373397
2,3-dimethyl-piperazine, piperazine, 2,3-dimethyl, 2,3-dimethylpiperazine, 2,3-dimethylpiperazine, cis + trans
2,3-dimethylpiperazine

Spezifikation

84468-52-0
C6H14N2
Air Sensitive
COWPTMLRSANSMQ-UHFFFAOYSA-N
2,3-dimethylpiperazine
522814
95%
81°C to 90°C
MFCD07373397
2,3-dimethyl-piperazine, piperazine, 2,3-dimethyl, 2,3-dimethylpiperazine, 2,3-dimethylpiperazine, cis + trans
CC1C(NCCN1)C
114.192
114.19
2,3-Dimethylpiperazine, cis + trans
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H228-H314-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate

RUO – Research Use Only