missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylquinoxaline, 97%, Thermo Scientific Chemicals

29.30 € - 98.30 €

Chemical Identifiers

CAS 2379-55-7
MDL Number MFCD00006728
InChI Key FKHNZQFCDGOQGV-UHFFFAOYSA-N
PubChem CID 16925
IUPAC Name 2,3-dimethylquinoxaline
SMILES CC1=NC2=CC=CC=C2N=C1C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10214050
View Documents
Thermo Scientific Acros
116780050
5 g
29.30 €
5g
Please to purchase this item. Need a web account? Register with us today!
10020860
View Documents
Thermo Scientific Acros
116780250
25 g
98.30 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2379-55-7
FKHNZQFCDGOQGV-UHFFFAOYSA-N
2,3-dimethylquinoxaline
MFCD00006728
16925
CC1=NC2=CC=CC=C2N=C1C

Specifications

2379-55-7
100.0
Beige
>100°C
97%
C10H10N2
23, 191
Solubility in water: insoluble
CC1=NC2=CC=CC=C2N=C1C
158.2
158.2
Crystalline Powder
96.0
102.0°C to 107.0°C
257.0°C to 259.0°C
Authentic
Glass bottle
MFCD00006728
quinoxaline, 2,3-dimethyl, aurora ka-3449, quinoxaline,3-dimethyl, 2,3-dimethyl-quinoxalin, 2,3-dimethyl-quinoxaline, acmc-1cg5s, quinoxaline,2,3-dimethyl, salor-int l498777-1ea, 5-23-07-00267 beilstein handbook reference, chembl67865
FKHNZQFCDGOQGV-UHFFFAOYSA-N
2,3-dimethylquinoxaline
16925
97%
2, 3-Dimethylquinoxaline, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear prote

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 219-162-0

missing translation for 'rtecsNumber' : VD1960000

missing translation for 'tsca' : TSCA

Product Identifier
  • 2,3-Dimethylquinoxaline

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.