missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Fluorophenyl)ethylamine, ≥97%, Thermo Scientific Chemicals

Product Code. 15482099
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 404-70-6
Molecular Formula C8H11FN
Molecular Weight (g/mol) 140.18
MDL Number MFCD00075376
InChI Key AUCVZEYHEFAWHO-UHFFFAOYSA-O
Synonym 3-fluorophenethylamine, 2-3-fluorophenyl ethylamine, 2-3-fluoro-phenyl-ethylamine, 2-3-fluorophenyl ethanamine, 2-3-fluorophenyl ethan-1-amine, benzeneethanamine, 3-fluoro, m-fluorophenethylamine, 3-flourophenethylamine, m-fluorophenylethylamine, 3-fluoro phenethylamine
PubChem CID 533928
IUPAC Name 2-(3-fluorophenyl)ethanamine
SMILES [NH3+]CCC1=CC=CC(F)=C1

Specifications

Density 1.066 g/mL
Boiling Point 87°C (15 mmHg)
Flash Point 83°C (181°F)
Refractive Index 1.509
Quantity 5 g
UN Number UN2735
Beilstein 3045789
Formula Weight 139.17
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 2-(3-Fluorophenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.