missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose is used as a pharmaceutical intermediate.
Solubility
Soluble in acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
2,3-O-Isopropylidene-alpha,beta-D-ribofuranose is used as a pharmaceutical intermediate.
Solubility
Soluble in acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
Quantity | 1 g |
Solubility Information | Soluble in acetone. |
Optical Rotation | −27° (c=1 in Chloroform) |
Formula Weight | 190.2 |
Chemical Name or Material | 2,3-O-Isopropylidene-alpha,beta-D-ribofuranose |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'