missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-(3-Piperidinyloxy)-N-propylacetamide
Chemical Identifiers
CAS | 902837-33-6 |
---|---|
Molecular Formula | C10H20N2O2 |
Molecular Weight (g/mol) | 200.282 |
MDL Number | MFCD08064279 |
InChI Key | HIPDESKOLOAVFF-UHFFFAOYSA-N |
Synonym | 2-piperidin-3-yloxy-n-propylacetamide, 2-3-piperidyloxy-n-propylacetamide, 2-piperidin-3-yl oxy-n-propylacetamide, 2-3-piperidinyloxy-n-propylacetamide |
PubChem CID | 43196816 |
IUPAC Name | 2-piperidin-3-yloxy-N-propylacetamide |
SMILES | CCCNC(=O)COC1CCCNC1 |
Chemical Identifiers
902837-33-6 | |
200.282 | |
HIPDESKOLOAVFF-UHFFFAOYSA-N | |
43196816 | |
CCCNC(=O)COC1CCCNC1 |
C10H20N2O2 | |
MFCD08064279 | |
2-piperidin-3-yloxy-n-propylacetamide, 2-3-piperidyloxy-n-propylacetamide, 2-piperidin-3-yl oxy-n-propylacetamide, 2-3-piperidinyloxy-n-propylacetamide | |
2-piperidin-3-yloxy-N-propylacetamide |
Specifications
902837-33-6 | |
MFCD08064279 | |
HIPDESKOLOAVFF-UHFFFAOYSA-N | |
2-piperidin-3-yloxy-N-propylacetamide | |
43196816 | |
2-(3-Piperidinyloxy)-N-propylacetamide |
C10H20N2O2 | |
2-piperidin-3-yloxy-n-propylacetamide, 2-3-piperidyloxy-n-propylacetamide, 2-piperidin-3-yl oxy-n-propylacetamide, 2-3-piperidinyloxy-n-propylacetamide | |
CCCNC(=O)COC1CCCNC1 | |
200.282 | |
200.28 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures