missing translation for 'onlineSavingsMsg'
Learn More

2,4,4-Trimethyl-1-pentene, 99%, Thermo Scientific Chemicals

Product Code. 10425755
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-39-1
Molecular Formula C8H16
Molecular Weight (g/mol) 112.21
MDL Number MFCD00008855
InChI Key FXNDIJDIPNCZQJ-UHFFFAOYSA-N
Synonym 2,4,4-trimethyl-1-pentene, diisobutylene, pentene, 2,4,4-trimethyl, 2,4,4-trimethylpentene, 1-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-4-pentene, unii-n69l73advf, 1-methyl-1-neopentylethylene, ccris 9103, 2,4,4-trimethylpentene-1
PubChem CID 7868
IUPAC Name 2,4,4-trimethylpent-1-ene
SMILES CC(=C)CC(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -101°C
Color Colorless
Density 0.7000g/mL
Boiling Point 101°C to 102°C
Flash Point -6°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)3CCH2C(CH3)=CH2
Refractive Index 1.4070 to 1.409
Quantity 500 mL
Specific Gravity 0.7
Solubility Information Solubility in water: immiscible with water
Formula Weight 112.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 4, 4-Trimethyl-1-pentene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'