missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetraaminopyrimidine sulfate, 97%

Product Code. 11483068
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11483068

Brand: Thermo Scientific Alfa Aesar A18788.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5392-28-9 | C4H10N6O4S | 238.22 g/mol

Used as an intermediate in the production of Methotrexate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an intermediate in the production of Methotrexate.

Solubility
Soluble in water and dimethyl sulfoxide.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5392-28-9
Molecular Formula C4H10N6O4S
Molecular Weight (g/mol) 238.22
MDL Number MFCD06408008
InChI Key MQEFDQWUCTUJCP-UHFFFAOYSA-N
Synonym 2,4,5,6-tetraaminopyrimidine sulfate, pyrimidinetetramine sulfate, pyrimidine-2,4,5,6-tetraamine sulfate, pyrimidine-2,4,5,6-tetramine; sulfuric acid, tetraaminopyrimidine sulfate, pyrimidinetetramine, sulfate 1:1, pyrimidinetetramine, sulfate, tetraaminopyrimidine sulphate, unii-53qqy0i99m, tetra-aminopyrimidine sulfate
PubChem CID 79358
IUPAC Name pyrimidine-2,4,5,6-tetramine;sulfuric acid
SMILES OS(O)(=O)=O.NC1=NC(N)=C(N)C(N)=N1

Specifications

Melting Point >300°C
Quantity 50 g
Beilstein 3785189
Solubility Information Soluble in water and dimethyl sulfoxide.
Formula Weight 238.23
Percent Purity 97%
Chemical Name or Material 2,4,5,6-Tetraaminopyrimidine sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.