missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetrachloropyrimidine, 97%

Product Code. p-196327 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15269826

Brand: Thermo Scientific Acros 333520010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1780-40-1 | C4Cl4N2 | 217.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1780-40-1
Molecular Formula C4Cl4N2
Molecular Weight (g/mol) 217.87
MDL Number MFCD00006062
InChI Key GVBHCMNXRKOJRH-UHFFFAOYSA-N
Synonym perchloropyrimidine, pyrimidine, tetrachloro, tetrachloropyrimidine, pyrimidine, 2,4,5,6-tetrachloro, 2,4,5,6-tetrachloro-1,3-pyrimidine, tetrachlorpyrimidin, tetrachloro-pyrimidin, acmc-1bsuq, 2,5,6-tetrachloropyrimidine, 5-23-05-00347 beilstein handbook reference
PubChem CID 15690
IUPAC Name 2,4,5,6-tetrachloropyrimidine
SMILES C1(=C(N=C(N=C1Cl)Cl)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 67°C to 68.5°C
Color White
Boiling Point 108°C to 112°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 217.87
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5,6-Tetrachloropyrimidine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.