missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,5-trifluoro-3-methoxybenzoyl chloride, Thermo Scientific™
49.90 € - 98.20 €
Chemical Identifiers
CAS | 112811-66-2 |
---|---|
Molecular Formula | C8H4ClF3O2 |
Molecular Weight (g/mol) | 224.563 |
InChI Key | JVQSZTJTWSUJCR-UHFFFAOYSA-N |
Synonym | 3-methoxy-2,4,5-trifluorobenzoyl chloride, pubchem8384, acmc-1c4v9, 2,4,5-trifluoro-3-methoxy-benzoyl chloride, benzoyl chloride,2,4,5-trifluoro-3-methoxy, benzoyl chloride, 2,4,5-trifluoro-3-methoxy, 2,4,5-trifluoro-3-methoxybenzoyl chloride, 2,4,5-tris fluoranyl-3-methoxy-benzoyl chloride |
PubChem CID | 2775281 |
IUPAC Name | 2,4,5-trifluoro-3-methoxybenzoyl chloride |
SMILES | COC1=C(C(=CC(=C1F)F)C(=O)Cl)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10756633
|
Thermo Scientific Maybridge
SB02064EA |
10 g |
49.90 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10163184
|
Thermo Scientific Maybridge
SB02064EB |
25 g |
98.20 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
112811-66-2 | |
224.563 | |
3-methoxy-2,4,5-trifluorobenzoyl chloride, pubchem8384, acmc-1c4v9, 2,4,5-trifluoro-3-methoxy-benzoyl chloride, benzoyl chloride,2,4,5-trifluoro-3-methoxy, benzoyl chloride, 2,4,5-trifluoro-3-methoxy, 2,4,5-trifluoro-3-methoxybenzoyl chloride, 2,4,5-tris fluoranyl-3-methoxy-benzoyl chloride | |
2,4,5-trifluoro-3-methoxybenzoyl chloride |
C8H4ClF3O2 | |
JVQSZTJTWSUJCR-UHFFFAOYSA-N | |
2775281 | |
COC1=C(C(=CC(=C1F)F)C(=O)Cl)F |
Specifications
112811-66-2 | |
C8H4ClF3O2 | |
JVQSZTJTWSUJCR-UHFFFAOYSA-N | |
2,4,5-trifluoro-3-methoxybenzoyl chloride | |
2775281 | |
97% |
95% | |
3-methoxy-2,4,5-trifluorobenzoyl chloride, pubchem8384, acmc-1c4v9, 2,4,5-trifluoro-3-methoxy-benzoyl chloride, benzoyl chloride,2,4,5-trifluoro-3-methoxy, benzoyl chloride, 2,4,5-trifluoro-3-methoxy, 2,4,5-trifluoro-3-methoxybenzoyl chloride, 2,4,5-tris fluoranyl-3-methoxy-benzoyl chloride | |
COC1=C(C(=CC(=C1F)F)C(=O)Cl)F | |
224.563 | |
224.56 | |
2, 4, 5-Trifluoro-3-methoxybenzoyl chloride |
Safety and Handling
GHS Signal Word: Danger