missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethylphenol, 99%, Thermo Scientific Chemicals

99.09 € - 347.73 €

Chemical Identifiers

CAS 496-78-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00020050
InChI Key VXSCPERJHPWROZ-UHFFFAOYSA-N
Synonym pseudocumenol, phenol, 2,4,5-trimethyl, 5-hydroxypseudocumene, 1-hydroxy-2,4,5-trimethylbenzene, unii-3pca9e425c, acmc-1bmse, 4-06-00-03247 beilstein handbook reference
PubChem CID 10335
IUPAC Name 2,4,5-trimethylphenol
SMILES CC1=CC(=C(C=C1C)O)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11375449
View Documents
Thermo Scientific Alfa Aesar
H50427.MD
250 mg
99.09 €
250mg
Please to purchase this item. Need a web account? Register with us today!
11385449
View Documents
Thermo Scientific Alfa Aesar
H50427.03
1 g
347.73 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

496-78-6
136.194
VXSCPERJHPWROZ-UHFFFAOYSA-N
10335
CC1=CC(=C(C=C1C)O)C
C9H12O
MFCD00020050
pseudocumenol, phenol, 2,4,5-trimethyl, 5-hydroxypseudocumene, 1-hydroxy-2,4,5-trimethylbenzene, unii-3pca9e425c, acmc-1bmse, 4-06-00-03247 beilstein handbook reference
2,4,5-trimethylphenol

Specifications

496-78-6
232°C
C9H12O
UN2430
VXSCPERJHPWROZ-UHFFFAOYSA-N
2,4,5-trimethylphenol
10335
99%
72°C
Phenol-like
MFCD00020050
pseudocumenol, phenol, 2,4,5-trimethyl, 5-hydroxypseudocumene, 1-hydroxy-2,4,5-trimethylbenzene, unii-3pca9e425c, acmc-1bmse, 4-06-00-03247 beilstein handbook reference
CC1=CC(=C(C=C1C)O)C
136.194
136.2
2,4,5-Trimethylphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.

missing translation for 'einecsNumber' : 207-832-5

missing translation for 'rtecsNumber' : SN2815000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 2,4,5-Trimethylphenol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : 2,4,5-Trimethylphenol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only