Learn More
2,4,6-Tribromo-3-hydroxybenzoic acid, 97%, Thermo Scientific Chemicals
23.10 € - 233.00 €
Chemical Identifiers
CAS | 14348-40-4 |
---|---|
Molecular Formula | C7H3Br3O3 |
Molecular Weight (g/mol) | 374.81 |
MDL Number | MFCD00055557 |
InChI Key | YDBHVMTTYXWHLI-UHFFFAOYSA-N |
Synonym | 3-hydroxy-2,4,6-tribromobenzoic acid, tbhba, benzoic acid, 2,4,6-tribromo-3-hydroxy, 2,4,6-tribromo-3-hydroxybenzoicacid, 2,4,6-thba, pubchem12541, acmc-209uep, ksc174o3h, 2,4,6-tribromo-3-hydroxy-benzoic acid, 2,4,6-tribromo-3-hydroxybenzoic acid |
PubChem CID | 151915 |
IUPAC Name | 2,4,6-tribromo-3-hydroxybenzoic acid |
SMILES | C1=C(C(=C(C(=C1Br)O)Br)C(=O)O)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11430248
|
Thermo Scientific Alfa Aesar
B23790.03 |
1 g | N/A | ||||||
11440248
|
Thermo Scientific Alfa Aesar
B23790.06 |
5 g |
74.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450248
|
Thermo Scientific Alfa Aesar
B23790.14 |
25 g |
233.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14348-40-4 | |
374.81 | |
YDBHVMTTYXWHLI-UHFFFAOYSA-N | |
151915 | |
C1=C(C(=C(C(=C1Br)O)Br)C(=O)O)Br |
C7H3Br3O3 | |
MFCD00055557 | |
3-hydroxy-2,4,6-tribromobenzoic acid, tbhba, benzoic acid, 2,4,6-tribromo-3-hydroxy, 2,4,6-tribromo-3-hydroxybenzoicacid, 2,4,6-thba, pubchem12541, acmc-209uep, ksc174o3h, 2,4,6-tribromo-3-hydroxy-benzoic acid, 2,4,6-tribromo-3-hydroxybenzoic acid | |
2,4,6-tribromo-3-hydroxybenzoic acid |
Specifications
14348-40-4 | |
C7H3Br3O3 | |
2844497 | |
YDBHVMTTYXWHLI-UHFFFAOYSA-N | |
2,4,6-tribromo-3-hydroxybenzoic acid | |
151915 | |
98% |
144°C to 149°C | |
MFCD00055557 | |
3-hydroxy-2,4,6-tribromobenzoic acid, tbhba, benzoic acid, 2,4,6-tribromo-3-hydroxy, 2,4,6-tribromo-3-hydroxybenzoicacid, 2,4,6-thba, pubchem12541, acmc-209uep, ksc174o3h, 2,4,6-tribromo-3-hydroxy-benzoic acid, 2,4,6-tribromo-3-hydroxybenzoic acid | |
C1=C(C(=C(C(=C1Br)O)Br)C(=O)O)Br | |
374.81 | |
374.81 | |
2,4,6-Tribromo-3-hydroxybenzoic acid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only