missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Tribromoaniline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 118°C to 122°C |
Density | 2.35 |
Boiling Point | 300°C |
Quantity | 100 g |
Beilstein | 2209258 |
Merck Index | 14,9610 |
Formula Weight | 329.83 |
Percent Purity | 98% |
Chemical Name or Material | 2,4,6-Tribromoaniline |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P23I050 | Certificate of Analysis | 12/04/2022 | A12453.36,A12453.22 |
10239637 | Certificate of Analysis | 12/04/2022 | A12453.36,A12453.22 |
5002F22P | Certificate of Analysis | 22/11/2021 | A12453.36,A12453.22 |
10213002 | Certificate of Analysis | 22/11/2021 | A12453.36,A12453.22 |
10205296 | Certificate of Analysis | 22/11/2021 | A12453.36,A12453.22 |
Safety and Handling


Product Identifier
- 2,4,6-Tribromoaniline
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 2,4,6-Tribromoaniline
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,4,6-Tribromoaniline, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction