missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromophenol, 98%

Product Code. 10091910 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10091910

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118-79-6 | C6H3Br3O | 330.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-79-6
Molecular Formula C6H3Br3O
Molecular Weight (g/mol) 330.80
MDL Number MFCD00002150
InChI Key BSWWXRFVMJHFBN-UHFFFAOYSA-N
Synonym tribromophenol, bromol, bromkal pur 3, xeroform, phenol, 2,4,6-tribromo, flammex 3bp, unii-ys6k3eu393, ccris 1658, great lakes ph-73, 5175-83-7 bismuth 3+ salt
PubChem CID 1483
ChEBI CHEBI:47696
IUPAC Name 2,4,6-tribromophenol
SMILES OC1=C(Br)C=C(Br)C=C1Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 89.0°C to 93.0°C
Color White to Brown
Boiling Point 244.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula Br3C6H2OH
Quantity 5 g
Beilstein 06,203
Fieser 02,424
Merck Index 15,9775
Solubility Information Solubility in water: 71mg/l (15°C). Other solubilities: soluble in acetone,diethylether,petroleum ether,,benzene,toluene,alcohols,chloroform,pyridine,and carbon tetrachloride
Sublimation Point 244°C
Formula Weight 330.79
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 4, 6-Tribromophenol, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4,6-Tribromophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.