missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trihydroxybenzoic acid monohydrate, 90+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
2,4,6-Trihydroxybenzoic acid monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4,6-Trihydroxybenzoic acid monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
2,4,6-Trihydroxybenzoic acid monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | ∼210°C (decomposition) |
Quantity | 25 g |
Beilstein | 2212148 |
Solubility Information | Soluble in water. |
Formula Weight | 188.14 (170.12 Anhydrous) |
Percent Purity | 94% |
Chemical Name or Material | 2,4,6-Trihydroxybenzoic acid monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'