missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trihydroxybenzoic acid monohydrate, 90+%

Product Code. 11459797
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11459797

Brand: Thermo Scientific Alfa Aesar B22768.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71989-93-0 | C7H8O6 | 188.135 g/mol

2,4,6-Trihydroxybenzoic acid monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trihydroxybenzoic acid monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71989-93-0
Molecular Formula C7H8O6
Molecular Weight (g/mol) 188.135
MDL Number MFCD00149095
InChI Key HWZIRFCGHAROOI-UHFFFAOYSA-N
Synonym 2,4,6-trihydroxybenzoic acid hydrate, 2,4,6-trihydroxybenzoic acid monohydrate, benzoic acid, 2,4,6-trihydroxy-, monohydrate, phloroglucinolcarboxylic acid, pubchem20337, acmc-209olb, 2,4,6-trihydroxybenzoicacidmonohydrate, 2,4,6-trihydroxybenzoic acid, hydrate, 2,4,6-trihydroxybenzoic acid monohydrate, technical t, 2,4,6-trihydroxybenzoic acid monohydrate, technical grade, predominantly 1,3,5-benzenetriol
PubChem CID 2723793
IUPAC Name 2,4,6-trihydroxybenzoic acid;hydrate
SMILES C1=C(C=C(C(=C1O)C(=O)O)O)O.O

Specifications

Melting Point ∼210°C (decomposition)
Quantity 100 g
Beilstein 2212148
Solubility Information Soluble in water.
Formula Weight 188.14 (170.12 Anhydrous)
Percent Purity 94%
Chemical Name or Material 2,4,6-Trihydroxybenzoic acid monohydrate
compliance-icons
Product Identifier
  • 2,4,6-Trihydroxybenzoic acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2,4,6-Trihydroxybenzoic acid monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.