Learn More
2,4,6-Triisopropylbenzenesulfonyl chloride, 98%
Beschreibung
2,4,6-Triisopropylbenzenesulfonyl chloride is used for the synthesis of glycerophospholipids as a condensing agent and in the analysis of phosphonolipids in egg yolk by HPLC/MS technique. It acts as a coupling reagent for the synthesis of oligonucleotides and as sulfonates of pyrimidine nucleosides. It also serves as a condensing agent in the synthesis of hydrogen-phosphonate diesters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,4,6-Triisopropylbenzenesulfonyl chloride is used for the synthesis of glycerophospholipids as a condensing agent and in the analysis of phosphonolipids in egg yolk by HPLC/MS technique. It acts as a coupling reagent for the synthesis of oligonucleotides and as sulfonates of pyrimidine nucleosides. It also serves as a condensing agent in the synthesis of hydrogen-phosphonate diesters.
Solubility
Soluble in chloroform.
Notes
Incompatible with strong bases.
Spezifikation
Spezifikation
| Melting Point | 94°C to 97°C |
| Quantity | 25 g |
| UN Number | UN3261 |
| Beilstein | 1218575 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in chloroform. |
| Formula Weight | 302.87 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,4,6-Triisopropylbenzenesulfonyl chloride |
Sicherheit und Handhabung
- 2,4,6-Triisopropylbenzenesulfonyl chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
- MIXTURE LIST-Contains : 2,4,6-Triisopropylbenzenesulfonyl chloride
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.