missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethyl-N-[4-(trifluoromethyl)benzyl]benzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15492076
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Reacts with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305849-16-5
Molecular Formula C17H18F3NO2S
Molecular Weight (g/mol) 357.391
MDL Number MFCD01652040
InChI Key FVZQXGUIEZGZJJ-UHFFFAOYSA-N
Synonym 2,4,6-trimethyl-n-4-trifluoromethyl benzyl benzenesulfonamide, 2,4,6-trimethyl-n-4-trifluoromethyl phenyl methyl benzenesulfonamide, 2,4,6-trimethyl-n-4-trifluoromethyl-benzyl-benzenesulfonamide
PubChem CID 3700146
IUPAC Name 2,4,6-trimethyl-N-[[4-(trifluoromethyl)phenyl]methyl]benzenesulfonamide
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)NCC2=CC=C(C=C2)C(F)(F)F)C

Specifications

Quantity 1 g
Beilstein 21328032
Formula Weight 357.39
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethyl-N-[4-(trifluoromethyl)benzyl]benzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'