missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

38.99 € - 433.16 €

Chemical Identifiers

CAS 5980-97-2
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.011
MDL Number MFCD00236060
InChI Key BZXQRXJJJUZZAJ-UHFFFAOYSA-N
Synonym mesitylboronic acid, 2,4,6-trimethylbenzeneboronic acid, 2-mesityleneboronic acid, 2,4,6-trimethylphenyl boronic acid, 2,4,6-trimethylphenyl boranediol, boronic acid, 2,4,6-trimethylphenyl, 2,4,6-trimethylboronic acid, boronic acid, b-2,4,6-trimethylphenyl, mesityl boronic acid, pubchem7895
PubChem CID 292184
IUPAC Name (2,4,6-trimethylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1C)C)C)(O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11481037
View Documents
Thermo Scientific Alfa Aesar
B24060.03
1 g
38.99 €
1g
Please to purchase this item. Need a web account? Register with us today!
11491037
View Documents
Thermo Scientific Alfa Aesar
B24060.06
5 g
104.52 €
5g
Please to purchase this item. Need a web account? Register with us today!
11401047
View Documents
Thermo Scientific Alfa Aesar
B24060.14
25 g
433.16 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5980-97-2
164.011
BZXQRXJJJUZZAJ-UHFFFAOYSA-N
292184
B(C1=C(C=C(C=C1C)C)C)(O)O
C9H13BO2
MFCD00236060
mesitylboronic acid, 2,4,6-trimethylbenzeneboronic acid, 2-mesityleneboronic acid, 2,4,6-trimethylphenyl boronic acid, 2,4,6-trimethylphenyl boranediol, boronic acid, 2,4,6-trimethylphenyl, 2,4,6-trimethylboronic acid, boronic acid, b-2,4,6-trimethylphenyl, mesityl boronic acid, pubchem7895
(2,4,6-trimethylphenyl)boronic acid

Specifications

5980-97-2
C9H13BO2
2831927
BZXQRXJJJUZZAJ-UHFFFAOYSA-N
(2,4,6-trimethylphenyl)boronic acid
292184
97%
∼120°C to 127°C
MFCD00236060
mesitylboronic acid, 2,4,6-trimethylbenzeneboronic acid, 2-mesityleneboronic acid, 2,4,6-trimethylphenyl boronic acid, 2,4,6-trimethylphenyl boranediol, boronic acid, 2,4,6-trimethylphenyl, 2,4,6-trimethylboronic acid, boronic acid, b-2,4,6-trimethylphenyl, mesityl boronic acid, pubchem7895
B(C1=C(C=C(C=C1C)C)C)(O)O
164.011
164.01
2,4,6-Trimethylbenzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.