Learn More
2,4,6-Trimethylbenzoyl chloride, 98+%, Thermo Scientific Chemicals
74.37 € - 955.00 €
Chemical Identifiers
CAS | 938-18-1 |
---|---|
Molecular Formula | C10H11ClO |
Molecular Weight (g/mol) | 182.647 |
MDL Number | MFCD00013650 |
InChI Key | UKRQMDIFLKHCRO-UHFFFAOYSA-N |
Synonym | mesitoyl chloride, benzoyl chloride, 2,4,6-trimethyl, 2,4,6-trimethylbenzoylchloride, 2,4,6-trimethyl-benzoyl chloride, pubchem13234, acmc-209rno, 2-mesitylenecarbonyl chloride, ksc491c5b, 2,4,6trimethylbenzoyl chloride, 2,4,6-trimethylbenzoyl cloride |
PubChem CID | 97038 |
IUPAC Name | 2,4,6-trimethylbenzoyl chloride |
SMILES | CC1=CC(=C(C(=C1)C)C(=O)Cl)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11417027
|
Thermo Scientific Alfa Aesar
A14407.09 |
10 g |
74.37 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11427027
|
Thermo Scientific Alfa Aesar
A14407.18 |
50 g |
270.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11437027
|
Thermo Scientific Alfa Aesar
A14407.30 |
250 g | N/A | ||||||
Description
2,4,6-Trimethylbenzoyl chloride is used, in the presence of pyridine, to protect OH groups as mesitoate esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4,6-Trimethylbenzoyl chloride is used, in the presence of pyridine, to protect OH groups as mesitoate esters.
Solubility
Reacts slowly with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. Air and moisture sensitive. Store under nitrogen. Incompatible with strong oxidizing agent
Chemical Identifiers
938-18-1 | |
182.647 | |
UKRQMDIFLKHCRO-UHFFFAOYSA-N | |
97038 | |
CC1=CC(=C(C(=C1)C)C(=O)Cl)C |
C10H11ClO | |
MFCD00013650 | |
mesitoyl chloride, benzoyl chloride, 2,4,6-trimethyl, 2,4,6-trimethylbenzoylchloride, 2,4,6-trimethyl-benzoyl chloride, pubchem13234, acmc-209rno, 2-mesitylenecarbonyl chloride, ksc491c5b, 2,4,6trimethylbenzoyl chloride, 2,4,6-trimethylbenzoyl cloride | |
2,4,6-trimethylbenzoyl chloride |
Specifications
938-18-1 | |
143°C to 146°C (60 mmHg) | |
C10H11ClO | |
MFCD00013650 | |
776108 | |
mesitoyl chloride, benzoyl chloride, 2,4,6-trimethyl, 2,4,6-trimethylbenzoylchloride, 2,4,6-trimethyl-benzoyl chloride, pubchem13234, acmc-209rno, 2-mesitylenecarbonyl chloride, ksc491c5b, 2,4,6trimethylbenzoyl chloride, 2,4,6-trimethylbenzoyl cloride | |
UKRQMDIFLKHCRO-UHFFFAOYSA-N | |
2,4,6-trimethylbenzoyl chloride | |
97038 | |
≥98% |
1.1 | |
>110°C (230°F) | |
1.529 | |
UN3265 | |
Moisture sensitive | |
Reacts slowly with water. | |
CC1=CC(=C(C(=C1)C)C(=O)Cl)C | |
182.647 | |
182.65 | |
2,4,6-Trimethylbenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 213-339-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 2,4,6-Trimethylbenzoyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
- MIXTURE LIST-Contains : 2,4,6-Trimethylbenzoyl chloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.