Learn More
2,4,6-Triphenylpyrylium tetrafluoroborate, 97%, Thermo Scientific Chemicals
Description
2,4,6-Triphenylpyrylium tetrafluoroborate is used as a sensitizer to carry out the photo oxidation of cathecol. It is also used in the preparation of three N-alkylpyridinium photosensitizers by reacting with (1R,2S)-(-)-norephedrine, (S)-(+)-2-(aminomethyl)pyrrolidine and (R)-(-)-1-cyclohexylethylamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,4,6-Triphenylpyrylium tetrafluoroborate is used as a sensitizer to carry out the photo oxidation of cathecol. It is also used in the preparation of three N-alkylpyridinium photosensitizers by reacting with (1R,2S)-(-)-norephedrine, (S)-(+)-2-(aminomethyl)pyrrolidine and (R)-(-)-1-cyclohexylethylamine.
Solubility
Insoluble in water, ethyl alcohol, diethyl ether. Soluble in trifluroacetic acid.
Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Melting Point | 248°C to 254°C |
Quantity | 5 g |
Beilstein | 3586533 |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water,ethyl alcohol,diethyl ether. Soluble in trifluroacetic acid. |
Formula Weight | 396.2 |
Percent Purity | 97% |
Chemical Name or Material | 2,4,6-Triphenylpyrylium tetrafluoroborate |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10244836 | Certificate of Analysis | 30/11/2022 | A18217.06,A18217.14,A18217.22 |
10241718 | Certificate of Analysis | 06/07/2022 | A18217.06,A18217.14,A18217.22 |
10241717 | Certificate of Analysis | 06/07/2022 | A18217.06,A18217.14,A18217.22 |
Y19E002 | Certificate of Analysis | 23/11/2021 | A18217.06,A18217.14,A18217.22 |
10210778 | Certificate of Analysis | 23/11/2021 | A18217.06,A18217.14,A18217.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.