missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Aminophenyl)ethylamine, 95%, Thermo Scientific Chemicals

Product Code. 10265530
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-00-9
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.2
MDL Number MFCD00008194
InChI Key LNPMZQXEPNWCMG-UHFFFAOYSA-N
Synonym 2-4-aminophenyl ethylamine, 4-2-aminoethyl aniline, 4-aminophenethylamine, 4-2-amino-ethyl-phenylamine, p-aminophenethylamine, benzeneethanamine, 4-amino, 4-2-aminoethyl benzenamine, 4-aminophenylethylamine, 4-2-aminoethyl phenylamine, 4-2-amino-ethyl-aniline
PubChem CID 72895
IUPAC Name 4-(2-aminoethyl)aniline
SMILES C1=CC(=CC=C1CCN)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 28°C
Color Brown-Yellow to Yellow
Density 1.0570g/mL
Boiling Point 103°C (0.3 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4CH2CH2NH2
Refractive Index 1.5905 to 1.5945
Quantity 5 g
Beilstein 13, I, 48
Specific Gravity 1.057
Formula Weight 136.2
Percent Purity 95%
Physical Form Viscous Liquid
Chemical Name or Material 2-(4-Aminophenyl)ethylamine, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(4-Aminophenyl)ethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.