missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Bromophenyl)ethylamine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
4-Bromophenethylamine was used in the synthesis of pyrazinoisoquinoline derivatives and N-2-(4-bromophenyl)ethyl chloroacetamide. It was also used in the synthesis of alkyl arylamino sufides employing elemental sulfur and various halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromophenethylamine was used in the synthesis of pyrazinoisoquinoline derivatives and N-2-(4-bromophenyl)ethyl chloroacetamide. It was also used in the synthesis of alkyl arylamino sufides employing elemental sulfur and various halides.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
4-Bromophenethylamine was used in the synthesis of pyrazinoisoquinoline derivatives and N-2-(4-bromophenyl)ethyl chloroacetamide. It was also used in the synthesis of alkyl arylamino sufides employing elemental sulfur and various halides.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
Color | Colorless |
Density | 1.290 g/mL |
Boiling Point | 63°C to 72°C (0.2 mmHg) |
Flash Point | 113°C (235°F) |
Refractive Index | 1.574 |
Quantity | 25 g |
UN Number | UN2735 |
Solubility Information | Slightly soluble in water. |
IUPAC Name | 2-(4-bromophenyl)ethanamine |
Formula Weight | 200.08 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'