missing translation for 'onlineSavingsMsg'
Learn More
2,4-Diaminophenol dihydrochloride, 98+%, Thermo Scientific Chemicals Product Code.: 11463278

2,4-Diaminophenol dihydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11463278
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11463278

Brand: Thermo Scientific Alfa Aesar A17697.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,4-Diaminophenol dihydrochloride (Amidol) is used in the dye manufacturing industries and as a color accelerator in photographic developers. It is also employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Diaminophenol dihydrochloride (Amidol) is used in the dye manufacturing industries and as a color accelerator in photographic developers. It is also employed as a pharmaceutical intermediate.

Solubility
Soluble in water; very faint turbidity [27.5 g/100 mL (15°C).]

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-09-7
Molecular Formula C6H10Cl2N2O
Molecular Weight (g/mol) 197.06
MDL Number MFCD00012979
InChI Key KQEIJFWAXDQUPR-UHFFFAOYSA-N
Synonym 2,4-diaminophenol dihydrochloride, amidol, dianol, acrol, 2,4-diaminophenol hcl, 2,4-diaminophenol.2hcl, phenol, 2,4-diamino-, dihydrochloride, 2,4-diaminophenol hydrochloride, diamidophenol hydrochloride, unii-t1iyy101nx
PubChem CID 8715
SMILES [H+].[H+].[Cl-].[Cl-].NC1=CC=C(O)C(N)=C1

Specifications

Melting Point ∼280°C (decomposition)
Quantity 25 g
UN Number UN2811
Beilstein 3913728
Merck Index 14,2983
Solubility Information Soluble in water; very faint turbidity [27.5g/100 mL (15°C).]
IUPAC Name 2,4-diaminophenol;dihydrochloride
Formula Weight 197.07
Percent Purity ≥98%
Chemical Name or Material 2,4-Diaminophenol dihydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242394 Certificate TypeCertificate of Analysis Date10/08/2022 Catalog NumberA17697.22,A17697.14
Lot Number10117658 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17697.22,A17697.14
Lot Number10212512 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17697.22,A17697.14
Lot NumberJ1405A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17697.22,A17697.14
Lot Number10167488 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17697.22,A17697.14
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Diaminophenol dihydrochloride, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.