missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-fluorobenzoic acid, 97%, Thermo Scientific™

Product Code. 11459567
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86522-89-6
Molecular Formula C7H2Cl2FO2
Molecular Weight (g/mol) 207.99
MDL Number MFCD00075281
InChI Key KZCWJHUTTSVCRO-UHFFFAOYSA-M
Synonym 2,4-dichloro-5-fluoro-benzoic acid, 2,4-dichloro-5-fluoro benzoic acid, benzoic acid, 2,4-dichloro-5-fluoro, pubchem1338, maybridge1_002392, acmc-209qa7, ksc448a5p, rarechem al be 1219, timtec-bb sbb003459, 2 4-dichloro-5-fluorobenzoic acid
PubChem CID 688138
IUPAC Name 2,4-dichloro-5-fluorobenzoic acid
SMILES [O-]C(=O)C1=CC(F)=C(Cl)C=C1Cl

Specifications

Melting Point 140°C to 142°C
Quantity 1 g
Formula Weight 209.01
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-5-fluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'