missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 13488388
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-84-0
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00002414
InChI Key ATCRIUVQKHMXSH-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dichloro, 2,4-dichlorobenzoesyre, 2,4 dichlorobenzoic acid, 2,4-dichloro-benzoic acid, 2,4-dichlorobenzoicacid, ccris 9472, furosemide impurity e, pubchem21313, acmc-209kqh, wln: qvr bg dg
PubChem CID 5787
ChEBI CHEBI:30748
IUPAC Name 2,4-dichlorobenzoic acid
SMILES C1=CC(=C(C=C1Cl)Cl)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 160.0°C to 164.0°C
Color White
Boiling Point >200.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula Cl2C6H3CO2H
Quantity 1 kg
Beilstein 09, 342
Solubility Information Solubility in water: 0.36g/L (15°C). Other solubilities: soluble in alcohol,ether and acetone,insoluble in heptane
Formula Weight 191.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 4-Dichlorobenzoic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dichlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'