Learn More
2,4-Dichlorobenzonitrile, 98%, Thermo Scientific Chemicals
39.00 € - 346.00 €
Chemical Identifiers
CAS | 6574-98-7 |
---|---|
Molecular Formula | C7H3Cl2N |
Molecular Weight (g/mol) | 172.008 |
MDL Number | MFCD00016373 |
InChI Key | GRUHREVRSOOQJG-UHFFFAOYSA-N |
Synonym | benzonitrile, 2,4-dichloro, 2,4-dichloro-benzonitrile, 2,4-dichlorobenzenecarbonitrile, acmc-209nrx, 2,4-dichloro benzonitrile, ksc352s3f |
PubChem CID | 81050 |
IUPAC Name | 2,4-dichlorobenzonitrile |
SMILES | C1=CC(=C(C=C1Cl)Cl)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11479352
|
Thermo Scientific Alfa Aesar
A10113.06 |
5 g |
39.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489352
|
Thermo Scientific Alfa Aesar
A10113.14 |
25 g |
113.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11499352
|
Thermo Scientific Alfa Aesar
A10113.22 |
100 g | N/A | ||||||
Description
Used as intermediate for the synthesis of agrochemicals, pharmaceuticals and chemical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as intermediate for the synthesis of agrochemicals, pharmaceuticals and chemical intermediates.
Solubility
Soluble in Methanol. Slightly soluble in water.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Chemical Identifiers
6574-98-7 | |
172.008 | |
GRUHREVRSOOQJG-UHFFFAOYSA-N | |
81050 | |
C1=CC(=C(C=C1Cl)Cl)C#N |
C7H3Cl2N | |
MFCD00016373 | |
benzonitrile, 2,4-dichloro, 2,4-dichloro-benzonitrile, 2,4-dichlorobenzenecarbonitrile, acmc-209nrx, 2,4-dichloro benzonitrile, ksc352s3f | |
2,4-dichlorobenzonitrile |
Specifications
6574-98-7 | |
C7H3Cl2N | |
UN3439 | |
benzonitrile, 2,4-dichloro, 2,4-dichloro-benzonitrile, 2,4-dichlorobenzenecarbonitrile, acmc-209nrx, 2,4-dichloro benzonitrile, ksc352s3f | |
GRUHREVRSOOQJG-UHFFFAOYSA-N | |
2,4-dichlorobenzonitrile | |
81050 | |
98% |
57°C to 61°C | |
MFCD00016373 | |
637738 | |
Soluble in Methanol. Slightly soluble in water. | |
C1=CC(=C(C=C1Cl)Cl)C#N | |
172.008 | |
172.01 | |
2,4-Dichlorobenzonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 229-493-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only