missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Difluoro-3'-methoxyacetophenone, 97%, Thermo Scientific™

Product Code. 11425800
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 373603-19-1
Molecular Formula C9H8F2O2
Molecular Weight (g/mol) 186.158
MDL Number MFCD02258906
InChI Key KTZOZLGEZVDGMS-UHFFFAOYSA-N
Synonym 2',4'-difluoro-3'-methoxyacetophenone, 2,4-difluoro-3-methoxy acetophenone, 2,4-difluoro-3-methoxyacetophenone, 1-2,4-difluoro-3-methoxyphenyl ethanone, 1-2,4-difluoro-3-methoxy-phenyl ethanone, 1-acetyl-2,4-difluoro-3-methoxybenzene, 1-2,4-difluoro-3-methoxyphenyl ethan-1-one
PubChem CID 3850760
IUPAC Name 1-(2,4-difluoro-3-methoxyphenyl)ethanone
SMILES CC(=O)C1=C(C(=C(C=C1)F)OC)F

Specifications

Refractive Index 1.494
Quantity 5 g
Formula Weight 186.15
Percent Purity 97%
Chemical Name or Material 2',4'-Difluoro-3'-methoxyacetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'