missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals
48.70 € - 441.00 €
Chemical Identifiers
CAS | 133730-34-4 |
---|---|
Molecular Formula | C8H11BO4 |
Molecular Weight (g/mol) | 181.982 |
MDL Number | MFCD01074590 |
InChI Key | SQTUYFKNCCBFRR-UHFFFAOYSA-N |
Synonym | 2,4-dimethoxybenzeneboronic acid, 2,4-dimethoxyphenyl boronic acid, 2,4-dimethoxyphenyl boranediol, boronic acid, 2,4-dimethoxyphenyl, 2,4-dimethoxybenzene boronic acid, boronic acid, b-2,4-dimethoxyphenyl, pubchem1823, acmc-209btd, ksc174i6j, 2,4-dimethoxyphenylboronicacid |
PubChem CID | 2734341 |
IUPAC Name | (2,4-dimethoxyphenyl)boronic acid |
SMILES | B(C1=C(C=C(C=C1)OC)OC)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11465807
|
Thermo Scientific Alfa Aesar
B24374.03 |
1 g |
48.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11475807
|
Thermo Scientific Alfa Aesar
B24374.06 |
5 g |
120.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485807
|
Thermo Scientific Alfa Aesar
B24374.14 |
25 g |
441.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
133730-34-4 | |
181.982 | |
SQTUYFKNCCBFRR-UHFFFAOYSA-N | |
2734341 | |
B(C1=C(C=C(C=C1)OC)OC)(O)O |
C8H11BO4 | |
MFCD01074590 | |
2,4-dimethoxybenzeneboronic acid, 2,4-dimethoxyphenyl boronic acid, 2,4-dimethoxyphenyl boranediol, boronic acid, 2,4-dimethoxyphenyl, 2,4-dimethoxybenzene boronic acid, boronic acid, b-2,4-dimethoxyphenyl, pubchem1823, acmc-209btd, ksc174i6j, 2,4-dimethoxyphenylboronicacid | |
(2,4-dimethoxyphenyl)boronic acid |
Specifications
133730-34-4 | |
C8H11BO4 | |
4744213 | |
SQTUYFKNCCBFRR-UHFFFAOYSA-N | |
(2,4-dimethoxyphenyl)boronic acid | |
2734341 | |
98% |
115°C to 120°C | |
MFCD01074590 | |
2,4-dimethoxybenzeneboronic acid, 2,4-dimethoxyphenyl boronic acid, 2,4-dimethoxyphenyl boranediol, boronic acid, 2,4-dimethoxyphenyl, 2,4-dimethoxybenzene boronic acid, boronic acid, b-2,4-dimethoxyphenyl, pubchem1823, acmc-209btd, ksc174i6j, 2,4-dimethoxyphenylboronicacid | |
B(C1=C(C=C(C=C1)OC)OC)(O)O | |
181.982 | |
181.98 | |
2,4-Dimethoxybenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only