missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11407717
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Yttrium and heavy lanthanide complexes is synthesised with 2, 4-dimethoxybenzoic acid and thermal and spectral features is studied. In the case of the electron-rich aromatic system 2,4-dimethoxybenzoic acid reduction of the intermediate HOBt ester is slow, and good.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Yttrium and heavy lanthanide complexes is synthesised with 2, 4-dimethoxybenzoic acid and thermal and spectral features is studied. In the case of the electron-rich aromatic system 2,4-dimethoxybenzoic acid reduction of the intermediate HOBt ester is slow, and good.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-52-1
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00002434
InChI Key GPVDHNVGGIAOQT-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dimethoxy, 2,4-dimethoxybenzoicacid, 2,4-dimethoxy benzoic acid, 2,4-dimethoxybenzoic, pubchem4949, rarechem al bo 0028, 2,4-dimethoxy-benzoicaci, 2,4 dimethoxybenzoic acid, 2.4-dimethoxybenzoic acid, dimethoxybenzoic-2,4 acid
PubChem CID 7052
IUPAC Name 2,4-dimethoxybenzoic acid
SMILES COC1=CC(=C(C=C1)C(=O)O)OC

Specifications

Melting Point 107°C to 111°C
Odor Odorless
Quantity 25 g
Beilstein 2210860
Solubility Information Slightly soluble in water.
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material 2,4-Dimethoxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'