missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzoyl chloride, 96%, Thermo Scientific™

Product Code. 11406130
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39828-35-8
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD00017527
InChI Key KVIUXRJCBBXEGJ-UHFFFAOYSA-N
Synonym 2,4-dimethoxybenzoylchloride, acmc-20aocn, 2, 4-dimethoxybenzoyl chloride, 2,4-dimethoxy benzoyl chloride, 2,4-dimethoxy-benzoyl chloride, benzoyl chloride,2,4-dimethoxy, 2,4-dimethoxybenzoic acid chloride, 2,4-bis methyloxy benzoyl chloride, 2,4-dimethoxy-1-benzenecarbonyl chloride, benzoyl chloride, 2,4-dimethoxy-9ci
PubChem CID 2734719
IUPAC Name 2,4-dimethoxybenzoyl chloride
SMILES COC1=CC(=C(C=C1)C(=O)Cl)OC

Specifications

Melting Point 57°C to 61°C
Quantity 25 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 200.62
Percent Purity 96%
Chemical Name or Material 2,4-Dimethoxybenzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'