missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-1,3-oxazole-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15585956
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2510-37-4
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.13
InChI Key JLSFKHJNJFXGAB-UHFFFAOYSA-N
Synonym 2,4-dimethyloxazole-5-carboxylic acid, 2,4-dimethyl-oxazole-5-carboxylic acid, dimethyl-1,3-oxazole-5-carboxylic acid, 2,4-dimethyl-1,3-oxazole-5-carboxylicacid, 2,4-dimethyl-5-oxazolecarboxylic acid, 5-oxazolecarboxylic acid, 2,4-dimethyl, pubchem17689, ukrorgsyn-bb bbv-156622, 2,4-dimethyl-oxazole-5-carboxylicacid, saltdata: free
PubChem CID 1132979
IUPAC Name 2,4-dimethyl-1,3-oxazole-5-carboxylic acid
SMILES CC1=C(OC(=N1)C)C(=O)O

Specifications

Melting Point 229.0°C to 231.0°C
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 141.13
Percent Purity 97%
Chemical Name or Material 2,4-Dimethyl-1,3-oxazole-5-carboxylic acid
compliance-icons
Product Identifier
  • 2,4-Dimethyl-1,3-oxazole-5-carboxylic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'