missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-2-pentenoic acid, predominantly trans, 99%

Product Code. p-7026614
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
This item is not returnable. View return policy

Product Code. 11455347

Brand: Thermo Scientific Alfa Aesar B22701.88

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 66634-97-7 | C7H12O2 | 128.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66634-97-7
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00209505
InChI Key DMHLGGQHOSTMJG-GQCTYLIASA-N
Synonym 2,4-dimethyl-2-pentenoic acid, unii-ki18c46dhr, 2e-2,4-dimethylpent-2-enoic acid, ki18c46dhr, 2,4-dimethyl-2-pentenoic acid, 2e, e-2,4-dimethyl-2-pentenoic acid, 2,4-dimethyl-2-pentenoic acid, predominantly trans, fema no. 3143, e-2,4-dimethylpent-2-enoic acid, 2-pentenoic acid, 2,4-dimethyl
PubChem CID 5362565
IUPAC Name (E)-2,4-dimethylpent-2-enoic acid
SMILES CC(C)C=C(C)C(=O)O

Specifications

Density 0.949
Refractive Index 1.454
Quantity 2.5 g
UN Number UN3265
Formula Weight 128.17
Percent Purity 99%
Chemical Name or Material 2,4-Dimethyl-2-pentenoic acid, predominantly trans

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.