missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-3-furoyl chloride, 90%, Thermo Scientific™

82.10 € - 239.30 €

Chemical Identifiers

CAS 15139-39-6
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.581
InChI Key NOEQXZGBSMGWCT-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-furoyl chloride, 2,4-dimethylfuran-3-carboxylic chloride, 3-furancarbonyl chloride, 2,4-dimethyl-9ci
PubChem CID 13019985
IUPAC Name 2,4-dimethylfuran-3-carbonyl chloride
SMILES CC1=COC(=C1C(=O)Cl)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10250700
View Documents Promotion Details
Thermo Scientific Maybridge
CC65802CB
250 mg Amber Glass Bottle
82.10 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10411440
View Documents Promotion Details
Thermo Scientific Maybridge
CC65802DA
1 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

15139-39-6
158.581
2,4-dimethyl-3-furoyl chloride, 2,4-dimethylfuran-3-carboxylic chloride, 3-furancarbonyl chloride, 2,4-dimethyl-9ci
2,4-dimethylfuran-3-carbonyl chloride
C7H7ClO2
NOEQXZGBSMGWCT-UHFFFAOYSA-N
13019985
CC1=COC(=C1C(=O)Cl)C

Specifications

15139-39-6
C7H7ClO2
NOEQXZGBSMGWCT-UHFFFAOYSA-N
2,4-dimethylfuran-3-carbonyl chloride
13019985
90%
90%
2,4-dimethyl-3-furoyl chloride, 2,4-dimethylfuran-3-carboxylic chloride, 3-furancarbonyl chloride, 2,4-dimethyl-9ci
CC1=COC(=C1C(=O)Cl)C
158.581
158.58
2, 4-Dimethyl-3-furoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications