missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Dimethylamino)phenyl]benzimidazole, 95%, Thermo Scientific™

Product Code. 15462419
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2562-71-2
Molecular Formula C15H15N3
Molecular Weight (g/mol) 237.31
MDL Number MFCD00441571
InChI Key ZKBBGUJBGLTNEK-UHFFFAOYSA-N
Synonym 2-p-n,n-dimethylaminophenyl-1h-benzoimidazole, 4-1h-benzimidazol-2-yl-n,n-dimethylaniline, 4-1h-benzoimidazol-2-yl-phenyl-dimethyl-amine, n-4-1h-benzimidazol-2-yl phenyl-n,n-dimethylamine, 4-benzimidazol-2-ylphenyl dimethylamine, 4-1h-1,3-benzodiazol-2-yl-n,n-dimethylaniline, cambridge id 5628722, 2-p-dimethylamino phenyl benzimidazole, 4-2-benzimidazolyl-n,n-dimethylaniline, 2-4-dimethylaminophenyl-1h-benzimidazole
PubChem CID 242786
IUPAC Name 4-(1H-benzimidazol-2-yl)-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)C1=NC2=CC=CC=C2N1

Specifications

Color Yellow
Quantity 250 mg
Beilstein 185491
Formula Weight 237.31
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-[4-(Dimethylamino)phenyl]benzimidazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.