missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzaldehyde, 99%, Thermo Scientific Chemicals

29.45 € - 325.62 €

Chemical Identifiers

CAS 15764-16-6
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00003340
InChI Key GISVICWQYMUPJF-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dimethyl, 2,4-dimethyl benzaldehyde, 2,4-xylylaldehyde, 2,4-dimethylbenzenecarboxaldehyde, unii-i06yu18h4a, 1-formyl-2,4-dimethylbenzene, 2,4-dimethyl-benzaldehyde, m-xylylaldehyde, fema no. 3427, 2,4-trimethyl benzaldehyde
PubChem CID 61814
IUPAC Name 2,4-dimethylbenzaldehyde
SMILES CC1=CC(=C(C=C1)C=O)C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10235690
View Documents
Thermo Scientific Acros
165790050
5 g Glass bottle
29.45 €
5g
Please to purchase this item. Need a web account? Register with us today!
10245690
View Documents
Thermo Scientific Acros
165790250
25 g Glass bottle
93.87 €
25g
Please to purchase this item. Need a web account? Register with us today!
10656924
View Documents
Thermo Scientific Acros
165791000
100 g Glass bottle
325.62 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15764-16-6
134.18
GISVICWQYMUPJF-UHFFFAOYSA-N
61814
CC1=CC(=C(C=C1)C=O)C
C9H10O
MFCD00003340
benzaldehyde, 2,4-dimethyl, 2,4-dimethyl benzaldehyde, 2,4-xylylaldehyde, 2,4-dimethylbenzenecarboxaldehyde, unii-i06yu18h4a, 1-formyl-2,4-dimethylbenzene, 2,4-dimethyl-benzaldehyde, m-xylylaldehyde, fema no. 3427, 2,4-trimethyl benzaldehyde
2,4-dimethylbenzaldehyde

Specifications

15764-16-6
100.0
Colorless
102.4°C to 103°C (14.0 mmHg)
Authentic
C9H10O
1.5482 to 1.5502
07, IV, 706
benzaldehyde, 2,4-dimethyl, 2,4-dimethyl benzaldehyde, 2,4-xylylaldehyde, 2,4-dimethylbenzenecarboxaldehyde, unii-i06yu18h4a, 1-formyl-2,4-dimethylbenzene, 2,4-dimethyl-benzaldehyde, m-xylylaldehyde, fema no. 3427, 2,4-trimethyl benzaldehyde
CC1=CC(=C(C=C1)C=O)C
134.18
134.18
Liquid
98.5
-9°C
0.9600g/mL
88°C
98.5% min. (GC)
(CH3)2C6H3CHO
MFCD00003340
0.96
GISVICWQYMUPJF-UHFFFAOYSA-N
2,4-dimethylbenzaldehyde
61814
99%
2, 4-Dimethylbenzaldehyde
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 239-856-7

Product Identifier
  • 2,4-Dimethylbenzaldehyde

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.