missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

61.90 € - 243.21 €

Chemical Identifiers

CAS 609-60-9
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.67
MDL Number MFCD00156143
InChI Key FREOGXBZEAMJQN-UHFFFAOYSA-N
Synonym 2,4-dimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4-dimethyl, 2,4-dimethyl-benzenesulfonyl chloride, 2,4-dimethylphenyl chlorosulfone, 2,4-dimethylbenzenesulfonylchloride, acmc-1avoq, ksc355s4b, tos-bb-1127, 2,4-dimethylphenylsulfonyl chloride, 2,4-dimethyl benzene sulfonyl chloride
PubChem CID 69109
SMILES CC1=CC=C(C(C)=C1)S(Cl)(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11479450
View Documents
Thermo Scientific Alfa Aesar
H50301.03
1 g
61.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
11489450
View Documents
Thermo Scientific Alfa Aesar
H50301.06
5 g
243.21 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

609-60-9
204.67
FREOGXBZEAMJQN-UHFFFAOYSA-N
69109
C8H9ClO2S
MFCD00156143
2,4-dimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4-dimethyl, 2,4-dimethyl-benzenesulfonyl chloride, 2,4-dimethylphenyl chlorosulfone, 2,4-dimethylbenzenesulfonylchloride, acmc-1avoq, ksc355s4b, tos-bb-1127, 2,4-dimethylphenylsulfonyl chloride, 2,4-dimethyl benzene sulfonyl chloride
CC1=CC=C(C(C)=C1)S(Cl)(=O)=O

Specifications

32°C to 33°C
Acrid
C8H9ClO2S
UN3261
FREOGXBZEAMJQN-UHFFFAOYSA-N
2,4-dimethylbenzene-1-sulfonyl chloride
69109
98%
609-60-9
1.555
MFCD00156143
2,4-dimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4-dimethyl, 2,4-dimethyl-benzenesulfonyl chloride, 2,4-dimethylphenyl chlorosulfone, 2,4-dimethylbenzenesulfonylchloride, acmc-1avoq, ksc355s4b, tos-bb-1127, 2,4-dimethylphenylsulfonyl chloride, 2,4-dimethyl benzene sulfonyl chloride
CC1=CC=C(C(C)=C1)S(Cl)(=O)=O
204.67
204.68
2,4-Dimethylbenzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.