missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylpyrrole, 97%, Thermo Scientific Chemicals

Product Code. 10105313
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-82-1
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.15
MDL Number MFCD00192088
InChI Key MFFMQGGZCLEMCI-UHFFFAOYSA-N
Synonym 2,4-dimethylpyrrole, 1h-pyrrole, 2,4-dimethyl, pyrrole, 2,4-dimethyl, unii-ynq49m599x, pubchem24003, 2,4-dimethyl-1h-pyrrol, acmc-209n5e, ksc493o6j, #
PubChem CID 39539
IUPAC Name 2,4-dimethyl-1H-pyrrole
SMILES CC1=CC(C)=CN1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown-Maroon to Orange-Red or Violet
Density 0.9240g/mL
Boiling Point 165.0°C to 167.0°C
Flash Point >109°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.4970
Quantity 1 g
Specific Gravity 0.924
Formula Weight 95.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dimethylpyrrole, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'