missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Diphenyl-1,3-thiazole-5-carboxylic acid, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 281.33 |
Percent Purity | 97% |
Chemical Name or Material | 2, 4-Diphenyl-1, 3-thiazole-5-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'