missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Ethoxyphenyl)ethanol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
Description
2-(4-Ethoxyphenyl)ethanol, is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(4-Ethoxyphenyl)ethanol, is used as a pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
2-(4-Ethoxyphenyl)ethanol, is used as a pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Specifications
Specifications
Melting Point | 40°C to 44°C |
Boiling Point | 138°C to 141°C (8 mmHg) |
Quantity | 2 g |
Beilstein | 3239377 |
Solubility Information | Soluble in water. |
Formula Weight | 166.22 |
Percent Purity | 98% |
Chemical Name or Material | 2-(4-Ethoxyphenyl)ethanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'